2,2,3,3-Tetrafluoropropyl-p-toluenesulfonate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,2,3,3-Tetrafluoropropyl 4-methylbenzenesulphonate

PC9037-100G PC9037-25G PC9037-5G
77418-516EA 344.36 USD
77418-516 77418-518 77418-520
2,2,3,3-Tetrafluoropropyl-p-toluenesulfonate
2,2,3,3-Tetrafluoropropyl-p-toluenesulfonate
Formula: C₁₀H₁₀F₄O₃S
MW: 286.25 g/mol
Boiling Pt: 134…136 °C/0.5 mmHg
Melting Pt: 13…16 °C
Density: 1.397 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00155960
CAS Number: 786-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR