4-{[5-(Trifluoromethyl)-2-pyridinyl]oxy}benzenecarbonitrile 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-((5-(Trifluoromethyl)pyridin-2-yl)oxy)benzonitrile

PC2103D-1G PC2103D-5G PC2103D-10G
77413-534EA 63.18 USD
77413-534 77413-536 77413-532
4-{[5-(Trifluoromethyl)-2-pyridinyl]oxy}benzenecarbonitrile 97%
4-{[5-(Trifluoromethyl)-2-pyridinyl]oxy}benzenecarbonitrile
Formula: C₁₃H₇F₃N₂O
MW: 264.21 g/mol
Melting Pt: 83…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00173780
CAS Number: 175277-01-7
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR