Nonafluoro-3,6-dioxaheptanoic acid 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC5519-1G PC5519-25G PC5519-5G
77416-778EA 27.5 USD
77416-778 77416-780 77416-782
Nonafluoro-3,6-dioxaheptanoic acid 98%
Nonafluoro-3,6-dioxaheptanoic acid
Formula: C₅HF₉O₄
Boiling Pt: 145…146 °C
Storage Temperature: Ambient
MDL Number: MFCD03095398
CAS Number: 151772-58-6
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR