5,5,5-Trifluoropentanoic acid 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC4455-1G PC4455-250MG PC4455-5G
77415-422EA 61.12 USD
77415-422 77415-424 77415-426
5,5,5-Trifluoropentanoic acid 97%
5,5,5-Trifluoropentanoic acid
Formula: C₅H₇F₃O₂
Boiling Pt: 94…95 °C/15 mmHg
Density: 1.293 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD02262192
CAS Number: 407-62-5
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR