N-[3-(Aminomethyl)phenyl]-N-methylamine 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-(Methylamino)benzylamine, 3-(Aminomethyl)-N-methylaniline

OR7679-1G OR7679-5G
77411-276EA 70.3 USD
77411-276 77411-278
N-[3-(Aminomethyl)phenyl]-N-methylamine 97%
N-[3-(Aminomethyl)phenyl]-N-methylamine
Formula: C₈H₁₂N₂
Boiling Pt: 148…152 °C
Storage Temperature: Ambient
MDL Number: MFCD04037894
CAS Number: 768343-60-8
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR