1-Methyl-1H-indole-4-carboxylic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR9222-5G OR9222-1G OR9222-250MG
77411-796EA 262.28 USD
77411-796 77411-780 77411-788
1-Methyl-1H-indole-4-carboxylic acid 95%
1-Methyl-1H-indole-4-carboxylic acid
Formula: C₁₀H₉NO₂
Melting Pt: 243…245 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08435874
CAS Number: 90924-06-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR