1-(2H-Pyrazol-3-yl)ethanone hydrochloride
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
5-Acetyl-1H-pyrazole hydrochloride, 1-(1H-Pyrazol-3-yl)ethanone hydrochloride
OR29765-1G
OR29765-250MG
77408-444EA
85.58
USD
77408-444
77408-446
1-(2H-Pyrazol-3-yl)ethanone hydrochloride
1-(2H-Pyrazol-3-yl)ethanone hydrochloride
Formula:
C₅H₇ClN₂O MW: 146.58 g/mol Melting Pt: 150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00204160 CAS Number: 175277-40-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...