1-Methyl-4-piperidin-4-ylpiperazine

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-Methyl-4-(piperidin-4-yl)piperazine

OR0885-1G OR0885-250MG OR0885-5G
77402-280EA 48.9 USD
77402-280 77402-282 77402-284
1-Methyl-4-piperidin-4-ylpiperazine
1-Methyl-4-piperidin-4-ylpiperazine
Formula: C₁₀H₂₁N₃
MW: 183.3 g/mol
Melting Pt: 53…56 °C
Storage Temperature: Ambient
MDL Number: MFCD03274729
CAS Number: 53617-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR