4H-Furo[3,2-b]pyrrole-5-carboxylic acid methylester 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Methyl 4H-furo[3,2-b]pyrrole-5-carboxylate

OR1443-10G OR1443-1G OR1443-5G
77404-960EA 819.12 USD
77404-960 77404-962 77404-964
4H-Furo[3,2-b]pyrrole-5-carboxylic acid methylester 95%
4H-Furo[3,2-b]pyrrole-5-carboxylic acid methylester
Formula: C₈H₇NO₃
MW: 165.15 g/mol
Melting Pt: 138…140 °C
Storage Temperature: Ambient
MDL Number: MFCD06797487
CAS Number: 77484-99-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR