Ethyl-1-tert-butyl-5-methyl-1H-pyrazole-3-carboxylate 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR30041-1G OR30041-250MG
77408-664EA 134.48 USD
77408-664 77408-666
Ethyl-1-tert-butyl-5-methyl-1H-pyrazole-3-carboxylate 95%
Ethyl-1-tert-butyl-5-methyl-1H-pyrazole-3-carboxylate
Formula: C₁₁H₁₈N₂O₂
MW: 210.28 g/mol
Boiling Pt: 130 °C/15 mmHg
Storage Temperature: Ambient
MDL Number: MFCD01313625
CAS Number: 519056-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR