1,4,6-Trimethyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR110357-1G
OR110357-5G
OR110357-10G
77403-274EA
77.44
USD
77403-274
77403-276
77403-272
1,4,6-Trimethyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile
1,4,6-Trimethyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile
Formula:
C₉H₁₀N₂O MW: 162.19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00829059 CAS Number: 64038-03-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...