2-Chloro-3,6-difluorophenylacetonitrile 98%

Supplier: APOLLO SCIENTIFIC

PC0020-1G PC0020-25G PC0020-5G
77412-344EA 54 USD
77412-344 77412-346 77412-348
2-Chloro-3,6-difluorophenylacetonitrile 98%
2-Chloro-3,6-difluorophenylacetonitrile
Formula: C₈H₄ClF₂N
Storage Temperature: Ambient
MDL Number: MFCD01631447
CAS Number: 261762-54-3
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR