2-Methyl-4-penten-2-ol 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR929828-1G OR929828-5G
77411-930EA 96.78 USD
77411-930 77411-932
2-Methyl-4-penten-2-ol 95%
2-Methyl-4-penten-2-ol
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 120 °C
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00021813
CAS Number: 624-97-5
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR