Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II) 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
(A-taPhos)2PdCl2, PdCl2(Amphos)2, Bis(di-tert-butyl(4-dimethylaminophenyl)phosphine)dichloropalladium(II), Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]dichloropalladium, Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II)
OR17048-10G
OR17048-1G
OR17048-5G
77405-920EA
513.48
USD
77405-920
77405-922
77405-924
Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II) 97%
Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II)
Formula:
C₃₂H₅₆Cl₂N₂P₂Pd MW: 708.08 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09265123 CAS Number: 887919-35-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...