3,5-Difluorophenyl isocyanate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1,3-Difluoro-5-isocyanatobenzene

PC0845-1G PC0845-25G PC0845-5G
77412-696EA 20.36 USD
77412-696 77412-698 77412-700
3,5-Difluorophenyl isocyanate
3,5-Difluorophenyl isocyanate

  • May form small amount of sediment.

Formula: C₇H₃F₂NO
MW: 155.104 g/mol
Boiling Pt: 159 °C
Density: 1.31 g/cm³ (25 °C)
Flash Pt: 54 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093693
CAS Number: 83594-83-6
UN: 3080
ADR: 6.1+3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR