5-Methyl-2-furanmethanol 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (5-Methylfuran-2-yl)methanol, 2-(Hydroxymethyl)-5-methylfuran

OR3029-1G OR3029-500MG
77408-844EA 52.98 USD
77408-844 77408-846
5-Methyl-2-furanmethanol 95%
5-Methyl-2-furanmethanol
Formula: C₆H₈O₂
MW: 112.128 g/mol
Density: 1.077 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00130134
CAS Number: 3857-25-8
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR