1-Boc-4-(2-hydroxyethyl)piperazine 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: tert-Butyl 4-(2-hydroxyethyl)piperazine-1-carboxylate, 4-(2-Hydroxyethyl)piperazine, N1-BOC protected

OR1550-25G OR1550-500MG OR1550-5G
77405-420EA 182.36 USD
77405-420 77405-422 77405-424
1-Boc-4-(2-hydroxyethyl)piperazine 98%
1-Boc-4-(2-hydroxyethyl)piperazine
Formula: C₁₁H₂₂N₂O₃
Boiling Pt: 114 °C/0.1 mmHg
Melting Pt: 39…43 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00728947
CAS Number: 77279-24-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR