1-Boc-3-iodoindole-5-carbonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR1717-1G OR1717-5G
77405-974EA 21.26 USD
77405-974 77405-976
1-Boc-3-iodoindole-5-carbonitrile 98%
1-Boc-3-iodoindole-5-carbonitrile
Formula: C₁₄H₁₃IN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD05864767
CAS Number: 864685-26-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR