5-Bromo-2-(difluoromethyl)pyridine 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC49144-1G PC49144-5G
77415-862EA 55.02 USD
77415-862 77415-864
5-Bromo-2-(difluoromethyl)pyridine 98%
5-Bromo-2-(difluoromethyl)pyridine
Formula: C₆H₄BrF₂N
Melting Pt: 35…40 °C
Flash Pt: 89.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11977429
CAS Number: 845827-13-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR