4-(Trifluoromethyl)styrene 96%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-(Trifluoromethyl)-4-vinylbenzene

PC8732-1G PC8732-5G
77418-440EA 29.54 USD
77418-440 77418-442
4-(Trifluoromethyl)styrene 96%
4-(Trifluoromethyl)styrene
Formula: C₉H₇F₃
MW: 172.15 g/mol
Boiling Pt: 65…66 °C/40 mmHg
Density: 1.165 g/cm³ (25 °C)
Flash Pt: 42 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075539
CAS Number: 402-50-6
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR