(2-Fluoro-4-nitrophenyl)methanol 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Fluoro-4-nitrobenzyl alcohol

PC57032-250MG PC57032-5G PC57032-1G
77416-954EA 24.26 USD
77416-954 77416-956 77416-952
(2-Fluoro-4-nitrophenyl)methanol 97%
(2-Fluoro-4-nitrophenyl)methanol
Formula: C₇H₆FNO₃
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD11110168
CAS Number: 660432-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR