2-(Perfluorobutyl)ethyl acrylate 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
PC5906D-1G
PC5906D-5G
77417-156EA
46.6
USD
77417-156
77417-158
77417-160
2-(Perfluorobutyl)ethyl acrylate 98%
2-(Perfluorobutyl)ethyl acrylate
Formula:
C₉H₇F₉O₂ Boiling Pt: 160 °C Melting Pt: –68 °C Density: 1.44 g/cm³ (25 °C) Flash Pt: 52 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236104 CAS Number: 52591-27-2 UN: 3272 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...