5-Methoxy-1-methyl-1H-indole-3-carbaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Methoxy-1-methylindole-3-carboxaldehyde

OR110442-1G OR110442-5G
77403-334EA 64.18 USD
77403-334 77403-336
5-Methoxy-1-methyl-1H-indole-3-carbaldehyde
5-Methoxy-1-methyl-1H-indole-3-carbaldehyde
Formula: C₁₁H₁₁NO₂
MW: 189.214 g/mol
Melting Pt: 126…129 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01578324
CAS Number: 39974-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR