4-Formylbenzeneboronic acid pinacol ester 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Formylphenylboronic acid pinacol ester, 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde, 4-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

OR14173-100G OR14173-25G OR14173-5G
77404-838EA 643.9 USD
77404-838 77404-840 77404-842
4-Formylbenzeneboronic acid pinacol ester 98%
4-Formylbenzeneboronic acid pinacol ester
Formula: C₁₃H₁₇BO₃
Melting Pt: 54…59 °C
Storage Temperature: Ambient
MDL Number: MFCD04972375
CAS Number: 128376-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR