2-Chloro-4-nitro-1H-imidazole

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Chloro-5-nitro-1H-imidazole

OR2577-1G OR2577-25G OR2577-5G
77407-646EA 21.28 USD
77407-646 77407-648 77407-650
2-Chloro-4-nitro-1H-imidazole
2-Chloro-4-nitro-1H-imidazole
Formula: C₃H₂ClN₃O₂
MW: 147.518 g/mol
Melting Pt: 214…216 °C
Storage Temperature: Ambient
MDL Number: MFCD03419295
CAS Number: 57531-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR