Vinyl methacrylate 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: Methacrylic Acid Vinyl Ester

OR920125-25G OR920125-5G
77411-736EA 75.4 USD
77411-736 77411-738
Vinyl methacrylate 98%
Vinyl methacrylate
Formula: C₆H₈O₂
MW: 112.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059329
CAS Number: 4245-37-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR