Tris(2,2,2-trifluoroethyl)borate 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Boric Acid Tris(2,2,2-trifluoroethyl) Ester, Tris(2,2,2-trifluoroethyl) borate

PC6808-1G PC6808-25G PC6808-5G
77417-536EA 28.52 USD
77417-536 77417-538 77417-540
Tris(2,2,2-trifluoroethyl)borate 97%
Tris(2,2,2-trifluoroethyl)borate
Formula: C₆H₆BF₉O₃
MW: 307.91 g/mol
Boiling Pt: 77 °C/200 mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD00093768
CAS Number: 659-18-7
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR