Propyl lactate 95%

Supplier: APOLLO SCIENTIFIC

Synonyms: Lactic Acid Propyl Ester

OR916585-25G OR916585-5G
77411-702EA 172.18 USD
77411-702 77411-704
Propyl lactate 95%
Propyl lactate
Formula: C₆H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00059661
CAS Number: 616-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR