N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate)

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-(Chloromethyl)-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane Bis(tetrafluoroborate), F-TEDA-BF4, 1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate)

PC3728K-100G PC3728K-250G PC3728K-25G
77414-852EA 96.78 USD
77414-852 77414-854 77414-856
N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate)
N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate)

Electrophilic fluorinating agent. Ask for technical literature(SelectfluorR - Registered trademark of Air Products).


Decomposes exothermally at >1000 °C.

Formula: C₇H₁₄B₂ClF₉N₂
Melting Pt: 260 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00142607
CAS Number: 140681-55-6
EINECS: 414-380-4
UN: 3088
ADR: 4.2,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR