N,N-Diethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-(Diethylamino)phenylboronic Acid Pinacol Ester, 2-[4-(Diethylamino)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR16404-1G OR16404-250MG OR16404-5G
77405-670EA 206.82 USD
77405-670 77405-672 77405-674
N,N-Diethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline 98%
N,N-Diethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Formula: C₁₆H₂₆BNO₂
MW: 275.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10000956
CAS Number: 920304-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR