N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine 99%

Supplier: APOLLO SCIENTIFIC

Synonyms: N,N′-Di(1-naphthyl)-N,N′-diphenyl-(1,1′-biphenyl)-4,4′-diamine, N4,N4'-Di(naphthalen-1-yl)-N4,N4'-diphenyl-[1,1'-biphenyl]-4,4'-diamine

OR51012-1G OR51012-5G
77410-150EA 21.4 USD
77410-150 77410-152
N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine 99%
N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine
Formula: C₄₄H₃₂N₂
MW: 588.75 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03093246
CAS Number: 123847-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR