N-(tert-Butoxycarbonyl)-1,4-diaminobutane 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: N-(4-Aminobutyl)carbamic Acid tert-Butyl Ester, N-(tert-Butoxycarbonyl)-1,4-butanediamine, NH2-C4-NH-Boc, N-Boc-1,4-diaminobutane, N-Boc-1,4-butanediamine, tert-Butyl N-(4-Aminobutyl)carbamate, tert-Butyl (4-aminobutyl)carbamate

OR307039-100G OR307039-25G OR307039-5G
77408-934EA 284.24 USD
77408-934 77408-936 77408-938
N-(tert-Butoxycarbonyl)-1,4-diaminobutane 97%
N-(tert-Butoxycarbonyl)-1,4-diaminobutane
Formula: C₉H₂₀N₂O₂
MW: 188.27 g/mol
Density: 0.984 g/cm³ (25 °C)
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00210019
CAS Number: 68076-36-8
UN: 2735
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR