Methyl heptafluoroisobutyrate 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Heptafluoroisobutyric Acid Methyl Ester

PC2122-1G PC2122-25G PC2122-5G
77413-540EA 38.72 USD
77413-540 77413-542 77413-544
Methyl heptafluoroisobutyrate 97%
Methyl heptafluoroisobutyrate
Formula: C₅H₃F₇O₂
Boiling Pt: 76…77 °C
Density: 1.49 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00633403
CAS Number: 680-05-7
UN: 3272
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR