Methyl-2-(trifluoromethyl)-3,3,3-trifluoropropionate 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-(Trifluoromethyl)-3,3,3-trifluoropropionic Acid Methyl Ester, Methyl 2,2-Bis(trifluoromethyl)acetate, 2,2-Bis(trifluoromethyl)acetic Acid Methyl Ester

PC5184E-1G PC5184E-25G PC5184E-5G
77416-460EA 75.4 USD
77416-460 77416-462 77416-464
Methyl-2-(trifluoromethyl)-3,3,3-trifluoropropionate 97%
Methyl-2-(trifluoromethyl)-3,3,3-trifluoropropionate
Formula: C₅H₄F₆O₂
MW: 210.08 g/mol
Boiling Pt: 90…91 °C
Density: 1.604 g/cm³ (25 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00054663
CAS Number: 360-54-3
UN: 3272
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR