L-Pipecolic acid 97%

Supplier: APOLLO SCIENTIFIC

OR480636-100G OR480636-25G
77410-028EA 251.64 USD
77410-028 77410-030
L-Pipecolic acid 97%
L-Pipecolic acid
Formula: C₆H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD00005981
CAS Number: 3105-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR