Ishikawa's reagent 25% in acetonitrile

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-(Diethylamino)-1,1,2,3,3,3-hexafluoropropane, Hexafluoropropene diethylamine, N,N-Diethyl-1,1,2,3,3,3-hexafluoropropylamine

PC32753-100G PC32753-25G PC32753-500G
77414-640EA 103.92 USD
77414-640 77414-642 77414-644
Ishikawa's reagent 25% in acetonitrile
Ishikawa's reagent
Formula: C₇H₁₁F₆N
Storage Temperature: Ambient
MDL Number: MFCD00054683
CAS Number: 309-88-6
UN: 2924
ADR: 3+8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR