ɑ-Acetamidocinnamic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Acetamido-3-phenylacrylic acid, α-Acetamidocinnamic acid

OR5000T-100G OR5000T-25G OR5000T-500G
77410-094EA 37.64 USD
77410-094 77410-096 77410-098
ɑ-Acetamidocinnamic acid
alpha-Acetamidocinnamic acid
Formula: C₁₁H₁₁NO₃
MW: 205.21 g/mol
Melting Pt: 188…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00188416
CAS Number: 5469-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR