4-Fluoro-ɑ-methylstyrene 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Fluoro-4-(1-methylethenyl)benzene, 2-(4-Fluorophenyl)-1-propene

PC3828-10G PC3828-1G PC3828-25G
77414-958EA 45.84 USD
77414-958 77414-960 77414-962
4-Fluoro-ɑ-methylstyrene 97%
4-Fluoro-alpha-methylstyrene
Formula: C₉H₉F
MW: 136.17 g/mol
Density: 1.03 g/cm³ (25 °C)
Flash Pt: 53 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00042297
CAS Number: 350-40-3
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR