4,4,5,5-Tetramethyl-2-vinyl-1,3,2-dioxaborolane 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Pinacol vinylboronate, Vinylboronic Acid Pinacol Ester

OR52135-100G OR52135-25G OR52135-5G
77410-216EA 481.9 USD
77410-216 77410-218 77410-220
4,4,5,5-Tetramethyl-2-vinyl-1,3,2-dioxaborolane 95%
4,4,5,5-Tetramethyl-2-vinyl-1,3,2-dioxaborolane
Formula: C₈H₁₅BO₂
MW: 154.02 g/mol
Boiling Pt: 35…40 °C/20 mmHg
Density: 0.908 g/cm³ (25 °C)
Flash Pt: 34 °C
Storage Temperature: Freezer
MDL Number: MFCD00192492
CAS Number: 75927-49-0
UN: 3272
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR