3-Vinylpyridine

Supplier: APOLLO SCIENTIFIC
Danger

OR8894-10G OR8894-1G OR8894-5G
77411-464EA 780.42 USD
77411-464 77411-466 77411-468
3-Vinylpyridine
3-Vinylpyridine
Formula: C₇H₇N
Boiling Pt: 85 °C/10 mmHg
Density: 0.98 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD02178019
CAS Number: 1121-55-7
UN: 3073
ADR: 6.1+3+8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR