3-Furaldehyde 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Furan-3-carbaldehyde

OR23028-10G OR23028-1G
77407-020EA 166.06 USD
77407-020 77407-022
3-Furaldehyde 98%
3-Furaldehyde
Formula: C₅H₄O₂
MW: 96.09 g/mol
Boiling Pt: 145 °C
Density: 1.15 g/cm³ (25 °C)
Flash Pt: 48 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010424
CAS Number: 498-60-2
UN: 1199
ADR: 6.1+3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR