3,5-Diisopropylpyrazole

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3,5-Diisopropyl-1H-pyrazole

OR921043-1G OR921043-25G OR921043-5G
77411-744EA 58.08 USD
77411-744 77411-746 77411-748
3,5-Diisopropylpyrazole
3,5-Diisopropylpyrazole
Formula: C₉H₁₆N₂
MW: 152.24 g/mol
Melting Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00671516
CAS Number: 17536-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR