3,3,5-Trimethylcyclohexylamine 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR937606-1G OR937606-5G
77412-016EA 54 USD
77412-016 77412-018
3,3,5-Trimethylcyclohexylamine 95%
3,3,5-Trimethylcyclohexylamine
Formula: C₉H₁₉N
MW: 141.26 g/mol
Boiling Pt: 177 °C
Density: 0.84 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00019379
CAS Number: 15901-42-5
UN: 2326
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR