2-Trifluoromethyl-2-propanol 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1,1,1-Trifluoro-2-methylpropan-2-ol

PC7750-1G PC7750-500MG
77418-162EA 23.4 USD
77418-162 77418-164
2-Trifluoromethyl-2-propanol 97%
2-Trifluoromethyl-2-propanol
Formula: C₄H₇F₃O
Boiling Pt: 80…81 °C
Density: 1.17 g/cm³ (25 °C)
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00014391
CAS Number: 507-52-8
EINECS: 208-075-3
UN: 1987
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR