2-Chloro-1,1,2-trifluoroethyl ethyl ether 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC2060-25G PC2060-5G
77413-496EA 54.52 USD
77413-496 77413-498
2-Chloro-1,1,2-trifluoroethyl ethyl ether 97%
2-Chloro-1,1,2-trifluoroethyl ethyl ether
Formula: C₄H₆ClF₃O
MW: 162.54 g/mol
Boiling Pt: 82 °C/630 mmHg
Density: 1.273 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00018849
CAS Number: 310-71-4
UN: 3271
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR