1-Trifluoromethyl-3,3-dimethyl-1,2-benziodoxole 96%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3,3-Dimethyl-1-(trifluoromethyl)-1,2-benziodoxole, Togni Reagent

PC56207-1G PC56207-5G
77416-822EA 54 USD
77416-822 77416-824
1-Trifluoromethyl-3,3-dimethyl-1,2-benziodoxole 96%
1-Trifluoromethyl-3,3-dimethyl-1,2-benziodoxole
Formula: C₁₀H₁₀F₃IO
MW: 330.09 g/mol
Melting Pt: 75…79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10567056
CAS Number: 887144-97-0
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR