1-Methyl-1H-imidazole-4,5-dicarboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4,5-Dicarboxy-1-methyl-1H-imidazole, 1-Methylimidazole-4,5-dicarboxylic Acid

OR10441-1G OR10441-5G
77402-718EA 75.4 USD
77402-718 77402-720
1-Methyl-1H-imidazole-4,5-dicarboxylic acid
1-Methyl-1H-imidazole-4,5-dicarboxylic acid
Formula: C₆H₆N₂O₄
MW: 170.12 g/mol
Melting Pt: 262 °C
Storage Temperature: Ambient
MDL Number: MFCD01552045
CAS Number: 19485-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR