11-Azido-3,6,9-trioxaundecan-1-amine 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-Amino-11-azido-3,6,9-trioxaundecane, Amino-PEG3-Azide, 11-Amino-3,6,9-trioxaundecanyl Azide, Azido-PEG3-Amine

OR8503T-1G OR8503T-5G
77411-404EA 151.8 USD
77411-404 77411-406
11-Azido-3,6,9-trioxaundecan-1-amine 97%
11-Azido-3,6,9-trioxaundecan-1-amine

Heterobifunctional PRG derivative. Contains a free aminethat can be conjugated to biological molecules directly by an amide linkage (or via the corresponding isothiocyanate).

Formula: C₈H₁₈N₄O₃
Boiling Pt: 198…202 °C
Density: 1.1 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00269874
CAS Number: 134179-38-7
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR