1-(2-Chloroethyl)-1H-hexahydroazepine hydrochloride 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
1-(2-Chloroethyl)hexamethyleneimine Hydrochloride, 1-(2-Chloroethyl)azepane hydrochloride
OR0989-100G
OR0989-25G
OR0989-5G
77402-372EA
198.66
USD
77402-372
77402-374
77402-376
1-(2-Chloroethyl)-1H-hexahydroazepine hydrochloride 98%
1-(2-Chloroethyl)-1H-hexahydroazepine hydrochloride
Formula:
C₈H₁₆ClN MW: 198.13 g/mol Melting Pt: 208…210 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012842 CAS Number: 26487-67-2 EINECS: 247-733-4 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...