(+)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride 99%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC5063J-1G
PC5063J-250MG
77416-384EA
182.36
USD
77416-384
77416-386
(+)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride 99%
(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride
Formula:
C₁₀H₈ClF₃O₂ Boiling Pt: 80 °C/14 mmHg Density: 1.34 g/cm³ (25 °C) Flash Pt: 89 °C Storage Temperature: Freezer |
MDL Number:
MFCD00067105 CAS Number: 20445-33-4 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...